Owl™ Dual Gel Vertical Electrophoresis System, Models P8, P9 and P10, Thermo Scientific
Supplier: Thermo Fisher Scientific
P8DS-1 P8DS-2 P8DS-3
27372-540EA 3841.5 CAD
27372-540 27372-542 27372-626 27372-628 27372-630 27372-634 27372-636
Owl™ Dual Gel Vertical Electrophoresis System, Models P8, P9 and P10, Thermo Scientific
Electrophoresis Systems Vertical Gel Electrophoresis Systems
For fast gel runs with flat, even banding
Self-contained design and clamping system simplify setup. Full-length, spring-loaded side clamps slide easily over the gel plate assembly, centering it on the upper buffer chamber. Molded gasket provides a leak-resistant seal. The 6.4mm (1/4") acrylic safety lid features an interlocking design to eliminate the risk of electrical shock while providing extra protection from low-level radioactive samples. Lid closes in one orientation only, ensuring that the (+) and (–) electrodes are not reversed. Three-electrode configuration improves fieldMore Product Information