Synthware Stopcock, Glass Plug, T-Bore
Supplier: Kemtech America
S310217 S311740 S312044
60005-174EA 166.35 CAD
60005-174 60005-176 60005-178
Synthware Stopcock, Glass Plug, T-Bore
Stopcocks
Stopcock with solid glass T-bore plug.
  • T-bore
  • Heavy wall borosilicate construction
  • All glass stopcock
Supplied with stainless steel constant pressure retention spring and clip.
Order Now
SPECIFICATIONS
Environmentally Preferable