Synthware Nitrogen/Argon Line Manifold with PTFE Stopcocks, Kemtech America
Supplier: Kemtech America
M211042 M211044 M531042 M541044
60004-556EA 711.59 CAD
60004-556 60004-558 60004-560 60004-566
Synthware Nitrogen/Argon Line Manifold with PTFE Stopcocks, Kemtech America
Manifolds
Manifolds feature a 14/20 outer joint at the center designed to fit a rubber septum
  • Heavy wall borosilicate construction
  • Teflon® stopcocks
  • 4 Ports
  • 14/20 joint at the center
The septum keeps syringes under an inert environment and aids in cutting down on the number used. The main tube has a 22 mm O.D., and is available with right, left, or both right and left side 10 mm hose connections. Delivery ports have 8 mm hose connections. All manifolds are equipped with four ports.
Order Now
SPECIFICATIONS
Environmentally Preferable