Synthware Separatory Funnels
Supplier: Kemtech America
F451010 F451025 F459025 F451050 F459050 F451100 F459100 F451125 F459125 F454125 F451250 F459250 F454250 F451010A F451025A F451050A F451100A F459100A F451125A F459125A F454125A F451250A F459250A F454250A
60003-846EA 257.54 CAD
60003-846 60003-850 60003-852 60003-854 60003-856 60003-858 60003-860 60003-862 60003-864 60003-866 60003-868 60003-870 60003-872 60003-874 60003-878 60003-882 60003-886 60003-888 60003-890 60003-892 60003-894 60003-896 60003-898 60003-900
Synthware Separatory Funnels
Funnels are equipped with a 2 mm PTFE stopcock.
  • Heavy wall Borosilicate construction
  • Ground glass joints
  • Teflon® stopcock
  • Available with Glass (A) or Teflon® Stopper
Available with a glass or PTFE stopper.