Synthware Peptide Synthesis Vessels with T-Bore PTFE Stopcock and Vacuum, Solid Phase
Supplier: Kemtech America
P140010M P140025M P140050M P140100M
60003-720EA 366.31 CAD
60003-720 60003-724 60003-728 60003-732
Synthware Peptide Synthesis Vessels with T-Bore PTFE Stopcock and Vacuum, Solid Phase
Reaction Vessels Reaction Vessels, Cylindrical Shape
Vessels feature an 8 mm O.D.
  • Teflon® Cap and stopcock
  • GL 14 Threaded cap
  • Medium fritted disc
  • 8 mm Sidearm and lower Ground glass joint
connection that is sealed on the inner joint for connection to an inner gas. Available with a fritted disc of coarse or medium porosity to support resin. The top of the vessel features a GL thread and a PTFE-lined screw cap. The PTFE stopcock has a 2 mm bore. The arm has an O.D. of 8 mm.