Synthware Peptide Synthesis Vessel with T-Bore PTFE Stopcock, Solid Phase, Kemtech America
Supplier: Kemtech America
P130010C P130050C P130050M P130100C P130100M
60003-700EA 362.08 CAD
60003-700 60003-708 60003-710 60003-712 60003-714
Synthware Peptide Synthesis Vessel with T-Bore PTFE Stopcock, Solid Phase, Kemtech America
Reaction Vessels Reaction Vessels, Cylindrical Shape
Vessels are available with a fritted disc of coarse or medium porosity to support resin.
  • Teflon® cap and stopcock
  • GL14 threaded cap
  • Medium or coarse fritted disc
  • 8 mm sidearm
The top of the vessel features a GL thread and a PTFE-lined screw cap. The PTFE stopcock has a 2 mm bore. The arm has an ext. Ø of 8 mm.