Synthware Peptide Synthesis Vessel, Solid Phase, Kemtech America
Supplier: Kemtech America
P120010C P120010M P120025C P120025M P120050C P120050M
60003-682EA 284.23 CAD
60003-682 60003-684 60003-686 60003-688 60003-690 60003-692
Synthware Peptide Synthesis Vessel, Solid Phase, Kemtech America
Reaction Vessels Reaction Vessels, Cylindrical Shape
Vessels are available with a fritted disc of coarse or medium porosity to support resin.
  • Teflon® Cap and stopcock
  • GL 14 Threaded Cap
  • Medium or coarse fritted disc
The top of the vessel features a GL thread and a PTFE-lined screw cap. The PTFE stopcock has a 2 mm bore. The arm has an O.D. of 8 mm.