KIMBLE® KIMAX® GL45 Media/Storage Bottles, Graduated, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
14395-100 14395-250 14395-500 14395-1000 14395-5000 14395-10000 14396-100 14396-250 14396-1000 14396-2000 14396-5000 14396-10000 14395-2000 14395-01
89000-926CS 286.23 CAD
89000-926 89000-928 89000-930 89000-932 89000-936 89000-938 89012-128 89012-130 89012-132 89012-134 89012-136 89012-138 89012-140 89000-934 10798-966
KIMBLE® KIMAX® GL45 Media/Storage Bottles, Graduated, DWK Life Sciences
Bottles Cell Culture Bottles
General-purpose bottles feature white enamel graduations and marking spots.
Wide, 30 mm Int.Ø openings are designed for easy filling. Replaceable, drip-free, clear polypropylene pour ring is incorporated into the bottle design. Bottles are available uncapped, or supplied with linerless GL45 polypropylene caps that are autoclavable to 140 °C (284 °F). Optional high-temperature caps and pour rings are autoclavable to 180 °C (356 °F).