Synthware Flasks with High Vacuum Valve
Supplier: Kemtech America
F490010 F490050 F490100 F490250 F490258 F490500 F49081L
60003-232EA 184.74 CAD
60003-232 60003-236 60003-238 60003-240 60003-242 60003-244 60003-250
Synthware Flasks with High Vacuum Valve
Reaction Vessels Reaction Vessels, Flask Shape
Flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
  • Hi Vacuum Valves
  • Externally threaded
  • Heavy wall borosilicate construction
  • Ground glass joints
Our externally threaded storage flask is designed for storage of air sensitive materials.