Synthware Storage Tube with Outer Joint, Kemtech America
Supplier: Kemtech America
F761425 F761450 F762450 F761100 F764100 F761108 F764108 F764258 F764358
60003-146EA 228.02 CAD
60003-146 60003-148 60003-152 60003-154 60003-158 60003-160 60003-164 60003-176 60003-188
Synthware Storage Tube with Outer Joint, Kemtech America
Flasks Nitrogen Flasks
Storage tubes are equipped with a high vacuum valve that features a PTFE O-ring at the tip.
  • Hi Vacuum valves
  • Externally Threaded
  • Heavy wall Borosilicate construction
  • Ground glass joints
Our externally threaded storage tube is designed for storage of air sensitive materials.