Synthware Distilling Bulbs, Kugelrohr, Kemtech America
Supplier: Kemtech America
D281005 D281010 D281015 D281025 D281050
60002-046EA 58.72 CAD
60002-046 60002-048 60002-050 60002-052 60002-054
Synthware Distilling Bulbs, Kugelrohr, Kemtech America
Distillation Components Distillation Receivers
Used in distillation of very small amount of distillate at lower temperature with high efficiency
  • Kugelrohr Design
  • Heavy wall borosilicate construction
  • Ground glass joints
[ST] joints size: 14/20.
Ordering information: Kugelrohr machine is required (not supplied). Vacuum adapter 60000-172 is sold separately.