KIMBLE® RAY-SORB® Amber GL 45 Media Bottle, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
14399-2000 14399-5000 14399-10000 14399-1000 14399-500 14399-100 14399-250
46000-180CS 224.09 CAD
46000-180 46000-182 46000-184 89000-966 89000-964 89230-852 89230-854
KIMBLE® RAY-SORB® Amber GL 45 Media Bottle, DWK Life Sciences
Bottles Cell Culture Bottles
RAY-SORB® process provides a shield to protect light-sensitive products, allowing less than 1% light transmission below 400nm and 5% transmission from 400 to 600 nm compared with untreated clear borosilicate glass.
Bottles feature enhanced 50 ml graduation intervals, drip-free pour ring, and a GL45 screw thread polypropylene cap.
Ordering information: Membrane caps are designed with a 0.2 µm PTFE membrane barrier to allow pressure equilibrium during low temperature autoclaving up to 140 °C (284 °F). The high-temperature caps are made of PBT with a PTFE-faced liner for good chemicalMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable