Synthware Reservoirs, Chromatography, Rodaviss® Joints, Kemtech America
Supplier: Kemtech America
C414250 C414500
60001-754EA 149.65 CAD
60001-754 60001-756
Synthware Reservoirs, Chromatography, Rodaviss® Joints, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Reservoir has a top 24/40 or 45/50 Rodaviss® outer joint for use with flow controller and the appropriate size lower inner joint.
  • Top 24/40 or 45/50 Rodaviss® outer joint
  • Open top cap, Viton® O-ring, and loosening ring included
Supplied with an open top cap, Viton® O-ring, and loosening ring.
Order Now
SPECIFICATIONS
Environmentally Preferable