Synthware Chromatography Columns with Spherical Joint, Fritted Disc, and PTFE Stopcock, Kemtech America
Supplier: Kemtech America
C195171C C195172C C195173C C195262C C195264C C195323C C195324C C195463C C195464C
60001-488EA 225.81 CAD
60001-488 60001-490 60001-492 60001-496 60001-500 60001-506 60001-508 60001-514 60001-516
Synthware Chromatography Columns with Spherical Joint, Fritted Disc, and PTFE Stopcock, Kemtech America
Chromatography Columns
Chromatography columns with stopcock and spherical joint with pinch clamp for a quick and tight connection under pressure.
  • Upper spherical joint
  • Heavy wall borosilicate construction
  • Ground glass joints
  • Teflon® stopcock
  • Coarse fritted disc
The coarse porosity fritted disc supports packing material.