Synthware Distillation Condensers with Removable Hose Connections, Kemtech America
Supplier: Kemtech America
C571120 C571200 C579110 C574200
60000-804EA 139.14 CAD
60000-804 60000-806 60000-808 60000-812
Synthware Distillation Condensers with Removable Hose Connections, Kemtech America
Condensers
Indentations at the bottom for supporting packing material.
  • Heavy wall borosilicate construction
  • Ground glass joints
  • Removable hose connections
Jacket permits the column to be used as a condenser.