Synthware Vacuum Filtration Adapters, Kemtech America
Supplier: Kemtech America
A841420 A842415 A842440
60000-688EA 91.86 CAD
60000-688 60000-692 60000-694
Synthware Vacuum Filtration Adapters, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Adapter is used with plain stem Buchner funnels and pluro-stopper.
  • With hose connection
  • Pluro-stoppers not included
Hose connection is 10 mm O.D. at the largest serration. Pluro-stoppers are sold separately.