Analog orbital shakers are designed for a wide range of applications, including cell cultures that require accurate and reproducible results.
- 15.9 kg (35 lbs.) capacity
- Microprocessor controls
- Continuous or timed operation
- Now with best-in-class, exclusive 5-year warranty
- Five-year limited warranty on parts and labor
They provide reproducible motion that is evenly distributed through the platform's entire surface
Operating features include variable speed microprocessor control that provides consistent, uniform mixing. An integrated analog timer can be set from 1 to 120 minutes. A powerful triple eccentric drive and brushless motor ensure reliable service in continuous duty and can handle loads up to 15.9 kg (35 lbs.). Shakers can run in cold rooms, incubators, and CO₂ environments from 0 to 40 °C (32 to 104 °F). To avoid splashing or sample loss, a ramping feature slowly increases speed to the desired set point.
Shakers can accommodate a variety of stainless steel platform options. Tier braces are constructed of stainless steel, allow stacking of same sized platforms, and provide a 25.4 cm (10) clearance between platforms. Adjustable platforms are equipped with 4 adjustable cushioned rollers to hold most glassware securely. Universal platforms are suitable for mounting flask clamps and test tube racks. The culture platforms with mat are ideal for slow speed applications that utilize low profile vessels such as Petri dishes or culture flasks. The separatory funnel platform holds up to three 500 ml to 2 L samples at once. Dedicated platforms feature pre-mounted PVC flask clamps for maximum utilization of space. Flask clamps are available in either stainless steel or PVC styles. Media bottle clamps and microplate clamps are constructed of stainless steel. Pivoting test tube racks feature stainless steel construction. Standard test tube racks feature a PVC-coated steel construction. Both rack styles come with hardware for easy attachment to platforms.
Additional applications include bacterial suspensions, staining/destaining, and general mixing.
Certifications: RoHS certified. TUV listed. 120 V units are CSA certified. 230 V units are CE marked.
Ordering information: Shaker is supplied with a nonskid rubber mat that fits the 33×27.9 cm (13×11) tray. Tray is suitable for holding Petri dishes and cell culture flasks. Unit also includes a 2.3 m (92") detachable 3-wire cord and plug. All platforms, clamps, and rack accessories are sold separately. 230 V, 50/60 Hz models are also available; contact your Avantor sales representative for more information.