PYREX® Potter-Elvehjem Tissue Grinder with Fluoropolymer Resin Pestles, Corning
Supplier: Corning
7725T-5 7725T-1 7725T-3 7725T-5X 7725T-8 7725T-17 7725T-45
89090-068CS 377.86 CAD
89090-068 22877-288 22877-290 22877-292 22877-294 22877-296 22877-298
PYREX® Potter-Elvehjem Tissue Grinder with Fluoropolymer Resin Pestles, Corning
Homogenizers Tissue Grinders
Homogenizers designed for preparing small samples for pathological and biochemical studies.
Glass pestle has fluoropolymer resin contact surface threaded onto a stainless steel shaft. Mortar has a tooled mouth and is not ground. Sample and buffer are forced through cylindrical portion of mortar as pestle is rotated downward.
Caution: Caution: If used with motor-driven device, attachment must contain some form of friction release clutch to prevent connective tissue or muscle from damaging apparatus. User should wear protective gloves of sufficient thickness in case glass components fail.