Fume Hood Safety Cabinets for Flammables, Justrite®
Supplier: Justrite
882407 882400 884807 884800 882427 882420 884827 883627 883620 883607 883600 883027 883020 883007 883000 882437 882430 882417 882410 882404 882414 882424 882434 883004 883024 883604 883624 884804 884824
97008-360EA 4929.76 CAD
97008-360 97008-362 97008-366 97008-368 97008-374 97008-376 97008-378 97008-386 97008-388 97008-392 97008-394 97008-428 97008-430 97008-440 97008-442 97008-468 97008-470 97008-476 97008-478 97014-334 97014-336 97014-338 97014-340 97014-342 97014-344 97014-346 97014-348 97014-350 97014-352
Fume Hood Safety Cabinets for Flammables, Justrite®
Furniture Storage Cabinets
Justrite™ laboratory safety cabinets are ideal for use with fume hoods.
  • Fully compliant, premium safety cabinets
  • Designed for safe storage and venting of solvents and flammable liquids
Cabinets easily fit under counters and lab benches and compliment existing cabinetry. A 10.2H×7.6D cm (4×3") kickplate conforms to most laboratory furniture. Fully welded, double-wall, 18-gauge steel cabinets have 3.8 cm (1¹/₂") of insulating air space. A 5.1 cm (2") sump captures spills. A galvanized steel shelf is adjustable on 5.7 cm (2¹/₄") centers and has a load capacity of 159 kg (350 lbs.). All cabinets have a three-point, self-adjusting lock and back wall vents with built-in flash arresters positioned at the lower left and top right. Adjustable leg levelers raise or lower the cabinet 1.6 cm (⁵/₈") at each corner. Durable powder-coat paint resists solvents, acids, and alkalies.
Certifications: Cabinets meet OSHA 29 CFR 1910.106 and NFPA Code 30 Section 4.3.3. FM approved. Self-closing door models are UFC compliant.
Ordering information: Ten-year limited manufacturer’s warranty. Cabinets are supplied with a 'Flammable' warning label affixed to the door. Additional colors are available upon request; contact your Avantor sales representative for details. See 97001-314 series for extra shelves and other accessories.