Black Phenolic Screw Caps, Rubber Liner, Qorpak®
Supplier: Qorpak
CAP-00469 CAP-00471 CAP-00475 CAP-00477 CAP-00481 CAP-00487 CAP-00489 CAP-00491 CAP-00493 CAP-00470
16214-998CS 955.54 CAD
16214-998 16215-017 16215-039 16215-040 16215-050 16215-061 16215-083 16215-094 16215-119 16215-130 16215-136 76539-334
Black Phenolic Screw Caps, Rubber Liner, Qorpak®
Closures Bottle Closures
Black phenolic caps with rubber liners are autoclavable
These black phenolic screw caps are made of phenolic resin.
This liner is FDA approved for food and pharmaceutical applications. Excellent properties of resilience, resistance to moisture vapor and relatively low gas transmission. Also widely used for packaging injectables. Great for culture work. They also provide the most consistent adherence to close dimensional tolerances.