Spacers for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Supplier: Thermo Fisher Scientific
ADJ1-C25 ADJ1-D25 P1-SC P2-SC P2-SD P7-SC P5-SD16
27372-004EA 37.97 CAD
27372-004 27372-008 27372-508 27372-560 27372-562 27372-601 27373-430
Spacers for Owl™ Multiple Gradient Gel Caster, Models P1 and P2, Thermo Scientific
Spacers
Spacers for 20 cm long plates are for protein gels only. Spacers for 45 cm long plates are for sequencing gels only.
Delivery information: Spacer set includes two side spacers and one bottom spacer.