Corrosive Storage Cabinets, SECURALL®
Supplier: Securall Cabinets
C160 C360 C224 C124 C130 C145 C330 C345
14226-892EA 7219.6 CAD
14226-892 14226-894 14227-648 15001-686 15001-688 15001-690 15001-692 15001-694
Corrosive Storage Cabinets, SECURALL®
Furniture Storage Cabinets
Cabinet features adjustable shelving and leveling legs, three-point latching system, 5.1 cm (2") vents with flash arrestors, flush-mounted locking handle, a sliding door, and a 5.1 cm (2") leakproof sill.
  • Large warning label reads 'CAUTION CORROSIVES'
  • Made with 18-gauge, galvanized steel
  • Double wall and a 3.8 cm (1¹/₂") air space
Shelves are equipped with removable corrosive-resistant polyethylene trays to contain spills and finished with two coats of corrosive-resistant blue epoxy enamel paint.
Made with 18-gauge, galvanized steel with a double wall and a 3.8cm (11/2") air space. Shelves are equipped with removable corrosive-resistant polyethylene trays to contain spills and finished with two coats of corrosive-resistant blue epoxy enamel paint. Large warning label reads “CAUTION CORROSIVES.”
Certifications: Cabinets comply with OSHA and NFPA Code 30 requirements. All cabinets, with the exception of 14227-648, are FM approved.
Caution: Not recommended for the storage of nitric or sulfuric acids.