Under-the-Counter Acid Cabinets, SciMatCo
Supplier: Scientific Materials
SC1648 SC1636 SC1630 SC1448 SC1436 SC1430
18500-024EA 8610.73 CAD
18500-024 18500-026 18500-028 18500-030 18500-032 18500-034 18500-036 18500-038
Under-the-Counter Acid Cabinets, SciMatCo
Furniture Storage Cabinets
Constructed of 2.5 cm (1") thick plywood, to be rust and corrosion proof
  • Finished with chemical resistant epoxy paint
  • Cabinets include padlock and two keys for security
Note: Dimensions include wood edge to wood edge height measurement of 87.6 cm (34½") without leveler.
Cabinets are made to fit under existing fume hoods or can be used as stand alone units. Cabinets feature a fixed position shelf, 5.1 cm (2") liquid-tight trough to contain spillage, and a wooden hinge that eliminates deterioration. Two styles are available. Partially lined with polypropylene on shelf and floor or fully lined with polypropylene on all interior walls, shelf, floor and top, except inside door surface.
Color: Blue