Glass Jugs, Amber
Supplier: Qorpak
259354 GLC-13300 259531 323201 323664
16087-018CS 208.88 CAD
16087-018 16194-105 93000-934 10770-318 10770-404 76474-074 76487-208 76487-210 76487-348 76500-060 76848-010 76848-014 77200-800
Glass Jugs, Amber
Bottles Carboys
Amber glass jugs with molded finger grips are the perfect storage solution for light sensitive products.
Amber glass protects against harmful UV rays.