Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1 and B2, Thermo Scientific
Supplier: Thermo Fisher Scientific
B1-BP B1A-BP B2-BP
27372-200EA 1428.27 CAD
27372-200 27372-202 27372-226 27372-228 27372-256 27372-258
Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1 and B2, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Mini-Gel systems have a small footprint and can run 5–34 samples (for 400 and 600mL systems) or 8–48 samples (for 800mL systems) on one gel
Leak-resistant, gasketed ultraviolet transmission trays allow tape-free casting in the buffer chamber. The trays have two comb positions and allow DNA bands to be viewed on a transilluminator without removing the gel. Optional multiload UVT trays have 12 slots for multiple sample-running, and allow samples to be loaded directly from a 96-well plate into an agarose gel. This design is based on the fixed spacing required to accommodate multichannel pipets (at least eight channels) and 96-well plates.
Heavy-duty, one-piece combs eliminate the need for comb assembly. The SuperSafe™ safety lid eliminates the risk of accidental shock. System is supplied with a 0.91m (3') safety power cord, attached.
The buffer chamber features lane visualization strips, which are silkscreened on the platform to provide a clear view of sample wells. Lane markers and fluorescent rulers provide accurate fragment size determination of DNA when compared to known standards. Durable acrylic construction and research-quality components give the buffer chamber a long life. Platinum electrodes and gold-plated banana plugs are corrosion-protected, and electrode placement ensures a uniform electric field.
Ordering information: Systems are supplied with manufacturer’s three-year warranty, and include a buffer chamber, a SuperSafe lid with attached power cords, an EasyCast gasketed UVT gel tray, and two 1.5mm thick combs. Mini-Gel Systems with gel dimensions measuring 7 x 8 cm contain one 6-well comb and one 10-well comb; systems with gel dimensions 9 x 11 cm contain one 10-well comb and one 14-well comb; and systems with gel dimensions 12 x 14 cm contain one 12-well comb and one 20-well comb. Each system is also available with built-in buffer exchange ports for use with circulating pumps.