VWR® Centrifugal Filters
Supplier: VWR International
CA82031-344 CA82031-352 CA82031-354 CA82031-360 CA82031-348 CA82031-356 82031-350 82031-360 82031-362 82031-346 82031-358 CA76196-216 CA76196-218 CA76196-220 CA76196-222 CA76196-224
CA82031-344PK 501.13 CAD
CA82031-344 CA82031-352 CA82031-354 CA82031-360 CA82031-348 CA82031-356 82031-350 82031-360 82031-362 82031-346 82031-358 CA76196-216 CA76196-218 CA76196-220 CA76196-222 CA76196-224
VWR® Centrifugal Filters
Filters Centrifugal Filters
Use to: Concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA; clean up labeling and PCR reactions; isolate DNA from agarose gel slices; buffer exchange; separate proteins, oligonucleotides, and RNA from acrylamide gels; clarify samples; or remove particulates.
  • Polypropylene housing
  • Membrane options available
Filter housings are constructed of polypropylene and are available with a polyethersulfone (PES) or nylon membrane.
Filters with nylon membrane are ideal for prefiltration clean-up of large agarose or acrylamide gel debris prior to further purification with an MWCO device.