JS-24.15 and JS-24.38 Swinging Bucket Rotors, Titanium Buckets, Beckman Coulter®
Supplier: Beckman Coulter
360743 362396
CABK360743EA 35385.62 CAD
CABK360743 CABK362396
JS-24.15 and JS-24.38 Swinging Bucket Rotors, Titanium Buckets, Beckman Coulter®
Centrifuges Rotors
The six-place, aluminum rotors with titanium swinging buckets are for use in Avanti™ J-26 XP and XPI and J6-MI centrifuge.
  • Ideal for Harvesting Bacteria, Processing Tissue Homogenates, Separating Cell Particulates, and More
  • Space for Six 15 mL Tubes or Six 38.5 mL Tubes
  • Titanium Swinging Buckets
  • Material: Aluminum
  • Six-Place Rotors
They are ideal for harvesting bacteria and subcellular particulates, processing tissue homogenates, and routine pelleting such as precipitates and phase separations.
They accommodate six 15mL tubes or six 38.5mL tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable