Orion™ Conductivity Standards, Thermo Scientific
Supplier: Thermo Fisher Scientific
01100610
01100510
01100710
14002-864PK
89.88
CAD
14002-864
14002-862
14002-866
Orion™ Conductivity Standards, Thermo Scientific
Standards
Conductivity Standards
These ready-to-use standards allow five conductivity levels to be easily measured in the field
Standards come in convenient 15mL pouches and certified against standards whose calibrations are traceable to the National Institute of Standards and Technology.
Ordering information: Store at room temperature.