Conductivity Standard, 10 µS/cm (10 µmho/cm) at 25°C 4.7 ppm as NaCl
Supplier: RICCA Chemical Company
R2246000-1A R2245000-4A R2245000-1A R2236000-500A R2237000-4A R2238000-4A R2243000-500A R2243000-4A R2237000-500A R2246000-4A R2238000-500A R2243000-1A R2245000-500A R2238000-1A R2246000-500A R2237000-1A R2236650-4A R2236000-4A R2236000-1A R2241000-1A R2241000-500A R2241000-4A
RC224132EA 128.39 CAD
RC224132 RC224332 RC2246-32 RC22411 RC22431 RC22381 RC2236-16 RC2236651 RC22451 RC2236-32 RC22461 RC224616 RC223716 RC22361 RC22371 RC223816 RC224316 RC223732 RC223832 RC224516 RC224532 RC224116
Conductivity Standard, 10 µS/cm (10 µmho/cm) at 25°C 4.7 ppm as NaCl
Standards Conductivity Standards
Calibration standards are individually calibrated and certified directly traceable to National Institute of Standards and Technology Electrolytic Conductivity Standard Reference Materials.
Contact your VWR representative for custom-prepared conductivity standards.
Each plastic container is labeled with the certified conductivity value, TDS concentration expressed as ppm sodium chloride (NaCl), and TDS expressed as equivalent 'Fresh Water Ion' concentrations when appropriate.