Teflo Air Sampling Membranes, PTFE, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
60146 R2PJ037 R2PJ047 R2PL047 R2PL037 60146 R2PJ047 R2PJ037 R2PI025 R2PI025
CA28139-109PK 737 CAD
CA28139-109 28139-131 CA28139-131 28150-400 87003-880 CA87003-880 CA28139-125 28139-129 28139-109 CA28139-129
Teflo Air Sampling Membranes, PTFE, Cytiva (Formerly Pall Lab)
Filters Membrane Filters
Strong, chemically resistant PTFE membranes have unique PMP (polymethylpentene) support ring for PM10 and PM2.5 dichotomous and other air sampling techniques.
  • Chemically resistant
  • PTFE
  • Increased durability
Membranes offer increased durability for hostile testing environments or acid aerosol monitoring.
Typical Aerosol Retention: 2 μm: 99.99%; 3 μm: 99.79%