Nalgene® Vacuum Chambers, Thermo Scientific
Supplier: Thermo Fisher Scientific
5305-0910 5305-1212 5305-0609
54929-062EA 913.17 CAD
54929-062 54929-084 54929-051
Nalgene® Vacuum Chambers, Thermo Scientific
Jars General Use Jars
Vacuum chamber consists of transparent jar, neoprene gasket and vacuum plate with adapter for 1/4" I.D tubing.
  • Transparent PC or Amber PEI bell jar
  • Gasketed white polycarbonate vacuum plate
Vacuum chamber 54929-051 has a polycarbonate jar and white polypropylene plate. It is intended only for applications using nonaggressive chemicals such as student demonstrations of vacuum procedures. Vacuum chambers 54929-062 and -084 have pale amber polyetherimide jars with excellent chemical resistance to acids, bases, aliphatic alcohols and hydrocarbons, andMore Product Information