Synthware Peptide Synthesis Vessels with Lower [ST] Joint and T-Bore PTFE Plug, Solid Phase
Supplier: Kemtech America
P150025M P150050C P150050M P150100M P150250M P150500M
60003-736EA 383.41 CAD
60003-736 60003-738 60003-740 60003-744 60003-748 60003-752
Synthware Peptide Synthesis Vessels with Lower [ST] Joint and T-Bore PTFE Plug, Solid Phase
Reaction Vessels Reaction Vessels, Cylindrical Shape
Solid phase peptide synthesis vessels feature 2 mm PTFE stopcocks and fritted glass supports for resin beads during operation.
  • Cap and stopcock included
  • Heavy wall borosilicate construction
  • Medium or coaorse porosity fritted disc available
The [ST] 24/40 inner drip joint can be assembled with other standard glassware. The vessel has an 8mm hose connection for vacuum applications. The stopcock is equipped with an 8mm O.D. sidearm for inert gas introduction. All vessels are supplied with PTFE-faced silicon-lined black nylon caps.