Pressure Tubes with Ace-Thred and Plunger Valve, Ace Glass
Supplier: Ace Glass
8648-60 8648-61 8648-62 8648-64 8648-65 8648-66 8648-68 8648-69 8648-70 8648-75 8648-76 8648-78 8648-79 8648-82 8648-83 8648-85 8648-86
89063-386EA 238.66 CAD
89063-386 89063-388 89063-390 89063-392 89063-394 89063-396 89063-398 89063-400 89063-402 89063-404 89063-406 89063-410 89063-412 89063-414 89063-416 89063-418 89063-420
Pressure Tubes with Ace-Thred and Plunger Valve, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall, borosilicate glass tubes are ideal for polymerization and analysis of halogens by thermal decomposition in the presence of lime
Available in a variety of lengths with either #7 or #15 Ace-Threds. Tubes are pressure rated to 150psi at 120°C (320°F).
The unique Ace-Thred design allows for easy closing with supplied plug and O-ring. Closures are reusable. Tubes are also equipped with bushing and plunger valve for purging. The closed-bottom plunger has a hole in the side that will open the tube to atmospheric conditions when properly positioned in relation to the O-ring seal.More Product Information