R2P™ and Modular R2P™ Roller Culture Apparatus, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W348888 W348889 W348887 W348886
14231-450EA 2250.89 CAD
14231-450 14231-452 74840-074 74840-072
R2P™ and Modular R2P™ Roller Culture Apparatus, Wheaton®, DWK Life Sciences
Roller Culture Apparatus
Designed to allow for the development and growth of cell culture from research to production quantities without having to change equipment and revalidate
Each deck holds five bottles. The roller apparatus offers the ability to start with as few as one deck (5 positions for bottles) and add up to 10 decks for a total of 55 bottle positions.
The Modular R2P™ systems feature a larger spacing between the decks than the standard R2P™ systems for inserting and removing roller bottles more easily without disturbing cells adhering to bottle walls.
Accommodates glass or plastic bottles 112More Product Information