Tabletop Horizontal Laminar Flow Workstations, Envirco®
Supplier: Envirco
10562 10564
VF-10562EA 25728.88 CAD
VF-10562 VF-10564
Tabletop Horizontal Laminar Flow Workstations, Envirco®
Fume Hoods and Enclosures Laminar Flow Cabinets
Self-contained multipurpose horizontal laminar flow workstations can be placed on an existing table, open bench, or support stand
The workstation exterior is constructed of welded aluminum with a corrosion-resistant polyurethane enamel finish, and it has removable clear acrylic side panels. The nonglare, plastic laminated work surface is easy to clean and stands 77.5cm (301/2") from the floor. A recessed fluorescent light fixture provides 150 foot-candles of illumination at the work surface.
Workstations utilize a direct-drive continuous-duty 1/3hp motor with sealed-for-life bearingsMore Product Information