Enviralab® Sterility Modules (ESM), Envirco®
Supplier: Envirco
10166 10170 10315
VF-10166EA 31687.18 CAD
VF-10166 VF-10170 VF-10315
Enviralab® Sterility Modules (ESM), Envirco®
Fume Hoods and Enclosures Laminar Flow Cabinets
These self-contained, laminar flow clean benches feature a modular design with an airflow pattern that offers the non-aspirating action of vertical flow while retaining the positive work protection of horizontal flow
Modules feature an all-stainless steel interior that includes fluorescent lighting and a shielded ultraviolet lamp to sterilize the work surface when the cabinet is not in use. For user protection, the ultraviolet lamp cannot be operated when the fluorescent light is in use. All modules also include a stainless steel IV rod, a covered duplex electrical outlet, a color-keyed airflow meter, and a solid-state speed control with RFI suppression for adjusting the airflow velocity.
Clean benches provide an ISO Class 5 (FED-STD-209E Class 100/M3.5) work environment by utilizing an aerosol-challenged HEPA filter rated 99.99% efficient at 0.3µm or larger. A 30% ASHRAE disposable prefilter is included to extend the life of the HEPA filter.
Certifications: CSA certified.
Ordering information: Supplied with a manufacturer's one-year limited warranty on mechanical parts and labor. Include a three-wire cord with plug. Optional support stand, lab service fixture, and additional duplex electrical outlets are available; contact your Avantor sales representative for more information.