Thermoplastic Duct Reducers, Labconco®
Supplier: Labconco
5605900 5606000 5606100 5630700
82006-934EA 521.4 CAD
82006-934 82006-936 82006-938 82006-940
Thermoplastic Duct Reducers, Labconco®
Ducting Supplies
PVC coupling type reducers are designed for connecting thermoplastic duct of different diameters
Compare your blower inlet size with your duct size to see if one is necessary.