Elbows, Labconco
Supplier: Labconco
4708700 4708800 5602100 7027300 7027400 5605100 5605200 4719000 4719100
30152-640EA 908.5 CAD
30152-640 30182-124 30182-136 30182-290 30182-326 82006-930 82006-932 30182-111 30182-144
Elbows, Labconco
Ducting Supplies
PVC elbows, both 45° and 90°, are compatible with thermoplastic duct
Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.