Eppendorf® 5430 Microcentrifuges
Supplier: Eppendorf
022620557 022620612 022620584 022620572 022620562 022620509 022620515 022620568 022620557 022620541 022620513 022620511 022620541 022620580 022620562 022620540 022620560 022620596 022620533 022620540 022620596 022620584 022620572 022620568 022620533 022620509 022620560 022620576 022620511 022620612
CA80094-182EA 8671.36 CAD
80094-172 CA80094-182 CA10030-956 CA80094-180 80094-176 CA10030-996 CA97027-850 97027-840 80094-178 89422-672 97027-838 CA97027-846 CA97027-844 CA97027-842 CA97027-840 490015-774 89422-674 75875-084 80094-182 10030-956 CA80094-172 CA80094-178 CA80094-176 10030-996 97027-850 97027-846 97027-844 97027-842 80094-180 CA97027-838
Eppendorf® 5430 Microcentrifuges
Centrifuges Microcentrifuges
The cross-over Eppendorf Centrifuge 5430 combines the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, it can accommodate rotors for microplates and 15/50 ml conical tubes as well as Eppendorf tubes and PCR strips.
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • Eppendorf QuickLock® - System for quickly opening and closing the rotor lid
Centrifuge 5430 combines the best features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument. The high-speed centrifuge offer centrifuge rotors for Eppendorf tubes and PCR strips, but also rotors for microplates and 15 or 50 ml conical tubes.
Additional Eppendorf QuickLock® centrifuge rotors are now available in the rotor program, including 48-place rotors (1.5 ml or 2.0 ml), a 16-place rotor for Eppendorf Tubes 5.0 ml and a swing-bucket rotor (24×1.5 ml or 2.0 ml). With its unmatched versatility, Centrifuge 5430 is the flagship product among the Eppendorf microcentrifuge portfolio.
Available as keypad or knob variant.
Certifications: UL Listed, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).