Target® Syringe Filters, PTFE, Thermo Scientific

Supplier: Thermo Scientific
F2500-4 F2502-3 F2500-13 F2513-4 F2500-3 F2513-3 F2504-3 F2504-4
66030-845PK 703.41 CAD
66030-845 66030-853 66064-393 66064-431 66030-841 66030-863 66030-874 66030-878
Target® Syringe Filters, PTFE, Thermo Scientific
Filters Syringe Filters
For use with nonaqueous-based and aggressive samples.

  • Resistant to all solvents, acids, and bases. Very low in extractables and low nonspecific binding
  • For use with aqueous samples, pre-wet the membrane with 3% MEOH and water, rinse with DI, and filter sample
  • The 4 mm filters can be used for volumes less than 2 ml
  • The 0.2 µm pore size filter can be used for 'cold' sterilization
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR