Target2™ Syringe Filters, Cellulose Acetate
Supplier: Thermo Scientific
F2504-16 F2504-15 F2500-15 F2500-16
66064-834CS 2193.31 CAD
66064-834 66064-836 66064-851 66064-854
Target2™ Syringe Filters, Cellulose Acetate
Filters Syringe Filters
Cellulose acetate membrane for biological or protein-based samples.
  • Low nonspecific binding with limited chemical resistance
  • Good flow rate characteristics
Use the 4 mm filters for limited volume samples up to 2 ml, and the 13 and 25 mm filters for volumes up to 100 ml. The 0.2 µm filters can be used for 'cold' sterilization.