Target® Syringe Filters, Regenerated Cellulose
Supplier: Thermo Scientific
F2500-7 F2504-7 F2504-8 F2513-7 F2500-8 F2513-8
66064-399PK 1228.78 CAD
66064-399 66064-412 66064-414 66064-435 66064-401 66064-437
Target® Syringe Filters, Regenerated Cellulose
Filters Syringe Filters
Hydrophilic regenerated cellulose membrane with low extractables and lowest nonspecific binding for biological applications or problem pharmaceutical samples.
  • Good chemical resistance
  • Housing material: Medical grade, solvent-resistant, virgin polypropylene
  • Pack of 100
Membrane improves sample yield when compared to polysulfone and cellulose acetate/nitrate membranes. The 4 mm filters are good for sample volumes up to 2 ml. The 0.2 µm pore size filter can also be used for 'cold' sterilization.