Target® Syringe Filters, Glass Microfiber, National Scientific™
Supplier: Thermo Scientific
F2500-20 F2500-19
66064-840CS 3608.68 CAD
66064-840 66064-842
Target® Syringe Filters, Glass Microfiber, National Scientific™
Filters Syringe Filters
Glass microfiber syringe filters are ideal for dissolution testing.
  • Polypropylene housing
  • Diameter: 30 mm
The glass microfiber filter is commonly used as a prefilter when samples are high in particulates because it helps to increase the load capacity of membranes. Solvent-resistant.