Galvanic Oxygen Sensors, WTW
Supplier: WTW
201545 201548 201425 G201533 201571 201549 201570 201547 201546
80085-410EA 4333.91 CAD
80085-410 80085-432 80085-420 80085-430 80085-424 80085-412 80085-422 80085-428 80085-426
Galvanic Oxygen Sensors, WTW
Electrodes Dissolved Oxygen Electrodes
These galvanic sensors are ready-to-use and are equipped with protective hoods for rough conditions.
CellOx* sensors are membrane-covered and waterproof. The membrane can be used for up to six months. Sensor also features high signal resolution, temperature compensation, rapid response, and membrane leakage monitoring. Includes a waterproof plug.
DurOx* oxygen sensors are membrane-covered and waterproof. Use for up to six months with only one electrolyte filling. Features temperature compensation and low approach flow. Includes a waterproof plug.
The StirrOx* self-stirring oxygen sensor simultaneously stirs and measures oxygen in Karlsruhe bottles and Winkler bottles. Sensor uses a constant approach flow for high reproducibility, consumes a very low amount of oxygen, only 0.008 µg h-1 (mg/l)-1, and features temperature compensation. Single-handed operation allows for rapid series measurements.
The SenTix* pH electrode features optimized membrane glass that guarantees quick measurement, even at low temperature.
Simple air calibration can be achieved by using a calibration vessel.
Delivery information: Sensors are supplied with an accessory case, spare parts, and maintenance supplies.