Conductivity Calibration Solutions, Beckman Coulter®

Supplier: BECKMAN COULTER CANADA LP CA

A58731
CABKA58730EA 115.39 CAD
CABKA58730 CABKA58731 CABKA58733 CABKA58732
Conductivity Calibration Solutions, Beckman Coulter®
Standards Conductivity Standards

Supplied in 946 mL (1 qt.) polyethylene bottles.


  • Accuracy: ±1% at 25°C (77°F)
  • Polyethylene Bottles
  • NIST-traceable Standards


Certifications: All conductivity standards are NIST traceable.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR