Conductivity Solutions, YSI
Supplier: XYLEM/YSI
060911 060660 065270 060907 065272 065274
23200-280CS 361.82 CAD
23200-280 23200-281 23202-220 23200-279 23202-285 23202-343
Conductivity Solutions, YSI
Standards Conductivity Standards
Use YSI Conductivity Solutions to calibrate conductivity cell and meter together as a system for maximum measurement accuracy.
Solutions are standardized against NIST Standard Reference Material potassium chloride (KCl). Each bottle includes calibration instruments and temperature correction information. Packaged in glass quart and plastic pint bottles.