Whatman™ Polydisc™ TF In-Line Filters
Supplier: Cytiva
6720-5002 6721-5010 6720-5001 6720-5045 6720-5001 6720-5045 6720-5002 6721-5010
28137-852PK 340.73 CAD
28137-852 28137-856 CA28137-850 CA28137-854 28137-850 28137-854 CA28137-852 CA28137-856
Whatman™ Polydisc™ TF In-Line Filters
Filters In-Line Filters
Whatman Polydisc TF In-Line Filters from Cytiva are lightweight devices with high chemical and solvent resistance that are particulraly suitable for protective vents or in-line filtration applications.
  • Chemically resistant hydrophobic PTFE membrane is well suited as a solvent filter for many different solutions
  • Filter can be autoclaved multiple times without losing integrity
  • Integrity-testable (bubble point or water breakthrough pressure in situ methods)
  • Lightweight to prevent tube collapsing
Filtering organic solvents and aggressive solutions calls for a chemically resistant, durable membrane. Cytiva's Polydisc TF features a PTFE membrane resistant to such compounds. The 1 µm membrane features a polypropylene prefilter for solvent filtration containing high particulate density in order to extend the useful life of the membrane.