Glass Microfiber Filter Paper Grade 151, Ahlstrom
Supplier: Ahlstrom
1510-0250 1510-0470 1510-0900 1510-1250 1510-1420 1510-1500 1510-0550 1510-0210 1510-0240 1510-0370 1510-0425 1510-0700 1510-1100
28496-174PK 265.98 CAD
28496-174 28496-176 28496-178 28496-180 28496-182 28496-184 38010-838 47749-578 47749-580 47749-582 47749-584 47749-586 47749-588
Glass Microfiber Filter Paper Grade 151, Ahlstrom
Filters Glass Fiber Filters
Ahlstrom Grade 151 is a binder-free microfiber glass filter designed to retain extremely fine particles.
It is recommended for use in the Toxicity Characteristic Leaching Procedure (TCLP) as described in EPA Method 1311.