VWR® Glass Fiber Filters
Supplier: VWR International
28297-289 28297-500 28297-504 28297-506 28297-508 28297-510 28297-974 28297-976 28297-978 28297-982 28297-984 28333-127 28333-129 28333-131 28333-133 28333-135 28333-137 28333-139 28333-125 CA28297-504 CA28297-506 CA28297-508 CA28333-133 CA28297-289 CA28297-500 CA28297-510 CA28297-974 CA28297-976 CA28297-978 CA28297-982 CA28297-984 CA28333-125 CA28333-127 CA28333-129 CA28333-131 CA28333-135 CA28333-137 CA28333-139
28297-289PK 178.51 CAD
28297-289 28297-500 28297-504 28297-506 28297-508 28297-510 28297-974 28297-976 28297-978 28297-982 28297-984 28333-127 28333-129 28333-131 28333-133 28333-135 28333-137 28333-139 28333-125 CA28297-504 CA28297-506 CA28297-508 CA28333-133 CA28297-289 CA28297-500 CA28297-510 CA28297-974 CA28297-976 CA28297-978 CA28297-982 CA28297-984 CA28333-125 CA28333-127 CA28333-129 CA28333-131 CA28333-135 CA28333-137 CA28333-139
VWR® Glass Fiber Filters
Filters Glass Fiber Filters
Binder-free VWR® glass fiber filters feature fast flow rates, high-loading capacity, wide thermal tolerance, and excellent precipitate retention.
  • Fast flow rates
  • High-loading capacity
  • Wide thermal tolerance
  • Excellent precipitate retention
Available in three grades: Grade 691 is an ideal filter for radioimmunoassay of weak beta emitters by scintillation counting, and for filtration of precipitated proteins, cells, and cell debris. Also used for gravimetric determination of airborne particulates. Grade 693 is recommended for use in dissolved and suspended solids testing in sanitary water analysis procedures. Grade 696 is used for monitoring specific pollutants (e.g., Hg in water), and in marine chemistry to filter particulate components in seawater. Also used in RIA procedures and for harvesting lymphocytes during incorporation trials.