Whatman™ Grade 597 and 597 ¹/₂ Qualitative Filter Papers
Supplier: Cytiva
10311847 10311851 10311844 10311812 10311845 10311853 10311862 10311809 10311811 10311822 10311809 10311862 10311841 10311808 10311810 10311810 10311841 10311822 10311808 10311811 10311814 10311814 10311807 10311820 10311897 10311854 10311856 10311843 10311852 10311804 10311887 10311842 10311804 10311807 10311820 10311842 10311843 10311852 10311854 10311856 10311887 10311897 10311847 10311851 10311844 10311812 10311845 10311853
14223-536PK 168.94 CAD
14223-536 14224-420 14224-424 14224-436 14224-640 74330-452 CA14224-434 CA14224-440 CA14224-442 CA74300-006 14224-440 14224-434 CA14223-590 CA11008-542 CA11008-544 11008-544 14223-590 74300-006 11008-542 14224-442 CA89233-650 89233-650 CA11012-660 CA11012-676 CA11012-682 CA11012-688 CA11012-690 CA11012-706 CA11012-744 CA11012-750 CA11012-754 CA11012-756 10034-664 10034-666 10034-668 10034-670 10034-672 10034-674 10034-676 10034-678 10034-680 10034-682 CA14223-536 CA14224-420 CA14224-424 CA14224-436 CA14224-640 CA74330-452
Whatman™ Grade 597 and 597 ¹/₂ Qualitative Filter Papers
Filters Filter Papers Qualitative Filter Paper
Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
  • Increased total loading capacity as more filter area is available
Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles. Cellulose materal (98% alpha cellulose content min.). Circles format.
A medium fast filter paper with medium to fine particle retention. Used for a wide variety of analytical routine applications in different industries like food testing (e.g., determination of fat content) or removal of carbon dioxide and turbidity from beverages (e.g., beer analysis).